Läs mer
2-(2-Aminoethyl)-3-(4-chlorophenyl)-3-hydroxyphthalimidine-d4 Hydrochloride (Mazindol Metabolite), TRC
Produktkod. 30325190
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30325190 25 mg
1 options
Produktkod. 30325190 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60891225MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(2-Aminoethyl)-3-(4-chlorophenyl)-3-hydroxyphthalimidine-d4 Hydrochloride (Mazindol Metabolite)
CAS 1276611-52-9
Molekylformel C16H12D4Cl2N2O2
Kvantitet 25 mg
Synonym 2-(2-Aminoethyl-d4)-3-(4-chlorophenyl)-2,3-dihydro-3-hydroxy-1H-isoindol-1-one Hydrochloride
InChI formel InChI=1S/C16H15ClN2O2.ClH/c17-12-7-5-11(6-8-12)16(21)14-4-2-1-3-13(14)15(20)19(16)10-9-18;/h1-8,21H,9-10,18H2;1H/i9D2,10D2;
LEDER C1=CC=C2C(=C1)C(=O)N(C([2H])([2H])C([2H])([2H])N)C2(C3=CC=C(C=C3)Cl)O.Cl
IUPAC-namn 2-(2-amino-1,1,2,2-tetradeuterioethyl)-3-(4-chlorophenyl)-3-hydroxyisoindol-1-one;hydrochloride
Molekylvikt (g/mol) 343.24