Läs mer
2-(2-Benzylphenoxy)-N-methylethanamine Hydrochloride, Mikromol™
Produktkod. 30051763
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051763 100 mg
30050311 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051763 Leverantör Mikromol Leverantörsnummer MM0770.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(2-Benzylphenoxy)-N-methylethanamine Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenyltoloxamine Related Compound A
CAS 75859-72-2
Analyt- eller komponentnamn Phenyltoloxamine Related Compound A
Frakt skick Room Temperature
Molekylformel C16 H19 N O . Cl H
Certifikat och efterlevnad ISO 17025
Synonym Ethanamine, N-methyl-2-[2-(phenylmethyl)phenoxy]-, hydrochloride (1:1), Ethanamine, N-methyl-2-[2-(phenylmethyl)phenoxy]-, hydrochloride (9CI), Ethylamine, N-methyl-2-(α-phenyl-o-tolyloxy)-, hydrochloride (5CI), Phenyltoloxamine Related Compound A as Hydrochloride
InChI formel InChI=1S/C16H19NO.ClH/c1-17-11-12-18-16-10-6-5-9-15(16)13-14-7-3-2-4-8-14;/h2-10,17H,11-13H2,1H3;1H
LEDER Cl.CNCCOc1ccccc1Cc2ccccc2
IUPAC-namn 2-(2-benzylphenoxy)-N-methylethanamine;hydrochloride
Molekylvikt (g/mol) 277.79
Kvantitet 100 mg
Formel vikt 277.1233 g/mol