Läs mer
2,2-Bis(4-hydroxyphenyl)propanol, TRC
Produktkod. 30012434
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012434 250 mg
30009944 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012434 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB447360250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 142648-65-5
Molekylformel C15 H16 O3
Molekylvikt (g/mol) 244.28
Synonym 4-Hydroxy-β-(4-hydroxyphenyl)-β-methyl-benzeneethanol
IUPAC-namn 4-[2-hydroxy-1-(4-hydroxyphenyl)-1-methyl-ethyl]phenol
LEDER CC(CO)(c1ccc(O)cc1)c2ccc(O)cc2
Specifikationer
Kemiskt namn eller material 2,2-Bis(4-hydroxyphenyl)propanol
Kvantitet 250 mg
InChI formel InChI=1S/C15H16O3/c1-15(10-16,11-2-6-13(17)7-3-11)12-4-8-14(18)9-5-12/h2-9,16-18H,10H2,1H3
Formel vikt 244.1099