missing translation for 'onlineSavingsMsg'
Läs mer

2-(2-Butylamino)ethanol, TRC

Produktkod. 30331570
Change view
Click to view available options
Kvantitet:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30331570 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331570 Supplier Toronto Research Chemicals Supplier No. TRCB6939455G

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-(2-Butylamino)ethanol
CAS 35265-04-4
Molekylformel C6H15NO
Kvantitet 5 g
Synonym 2-(sec-Butylamino)-ethanol, 2-[(1-Methylpropyl)amino]ethanol, Alpamine N 41, N-(2-Hydroxyethyl)-N-(butan-2-yl)amine, NSC 128123, sec-Butylmonoethanolamine
InChI formel InChI=1S/C6H15NO/c1-3-6(2)7-4-5-8/h6-8H,3-5H2,1-2H3
LEDER CCC(C)NCCO
IUPAC-namn 2-(butan-2-ylamino)ethanol
Molekylvikt (g/mol) 117.19
Formel vikt 117.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.