Läs mer
2-(2-Chloro-1-iminoethyl)-1,3,5-benzenetriol Hydrochloride, TRC
Produktkod. 30361890
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30361890 5 g
1 options
Produktkod. 30361890 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5961455G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(2-Chloro-1-iminoethyl)-1,3,5-benzenetriol Hydrochloride
CAS 468721-11-1
Molekylformel C8H8ClNO3 . HCll
Kvantitet 5 g
Synonym 1,3,5-Benzenetriol, 2-(2-chloro-1-iminoethyl)-, hydrochloride (1:1) (ACI), 1,3,5-Benzenetriol, 2-(2-chloro-1-iminoethyl)-, hydrochloride (9CI), 2-(2-Chloro-1-iminoethyl)-1,3,5-benzenetriol hydrochloride
InChI formel InChI=1S/C8H8ClNO3.ClH/c9-3-5(10)8-6(12)1-4(11)2-7(8)13;/h1-2,10-13H,3H2;1H
LEDER Cl.Oc1cc(O)c(C(=N)CCl)c(O)c1
IUPAC-namn 2-(2-chloroethanimidoyl)benzene-1,3,5-triol;hydrochloride
Molekylvikt (g/mol) 238.06
Formel vikt 236.9959