missing translation for 'onlineSavingsMsg'
Läs mer

2-(2-Chloro-6-fluorobenzylthio)aniline, TRC

Product Code. 30363948
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30363948 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30363948 Supplier Toronto Research Chemicals Supplier No. TRCC349080100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-(2-Chloro-6-fluorobenzylthio)aniline
CAS 646989-63-1
Molekylformel C13H11ClFNS
Kvantitet 100 mg
InChI formel InChI=1S/C13H11ClFNS/c14-10-4-3-5-11(15)9(10)8-17-13-7-2-1-6-12(13)16/h1-7H,8,16H2
LEDER Nc1ccccc1SCc2c(F)cccc2Cl
IUPAC-namn 2-[(2-chloro-6-fluorophenyl)methylsulfanyl]aniline
Molekylvikt (g/mol) 267.75
Formel vikt 267.0285
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.