Läs mer
2-(2-Chlorobenzoyloxy)acetophenone, TRC
Produktkod. 30361807
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361807 250 mg
30350872 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30361807 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC463410250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(2-Chlorobenzoyloxy)acetophenone
CAS 84634-61-7
Molekylformel C15H11ClO3
Kvantitet 250 mg
Synonym 2-Acetylphenyl 2-Chlorobenzoate
InChI formel InChI=1S/C15H11ClO3/c1-10(17)11-6-3-5-9-14(11)19-15(18)12-7-2-4-8-13(12)16/h2-9H,1H3
LEDER CC(=O)c1ccccc1OC(=O)c2ccccc2Cl
IUPAC-namn (2-acetylphenyl) 2-chlorobenzoate
Molekylvikt (g/mol) 274.7
Formel vikt 274.0397