Läs mer
2-(2-Chloroethoxy)acetyl Chloride, TRC
Produktkod. 30364320
Change view
Click to view available options
Kvantitet:
1 g
Product Code. Kvantitet
30364320 1 g
1 options
Product Code. 30364320 Supplier Toronto Research Chemicals Supplier No. TRCC3657151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 2-(2-Chloroethoxy)acetyl Chloride
CAS 39229-33-9
Molekylformel C4 H6 Cl2 O2
Kvantitet 1 g
InChI formel InChI=1S/C4H6Cl2O2/c5-1-2-8-3-4(6)7/h1-3H2
LEDER ClCCOCC(=O)Cl
IUPAC-namn 2-(2-chloroethoxy)acetyl chloride
Molekylvikt (g/mol) 157
Formel vikt 155.9745