missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chlorophenyl)benzimidazole, 98%, Thermo Scientific™

Catalog Number 11351997
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3574-96-7
C13H9ClN2
228.679
MFCD00051632
OILIYWFQRJOPAI-UHFFFAOYSA-N
chlorfenazole, 2-2-chlorophenyl benzimidazole, 2-2-chlorophenyl-1h-benzimidazole, chlorfenazole iso, 1h-benzimidazole, 2-2-chlorophenyl, 2-2-chlorophenyl-1h-benzoimidazole, 2-2-chlorophenyl-1h-benzo d imidazole, unii-rt668w2svp, 2-o-chlorophenyl benzimidazole, 2-2-chlorophenyl-1h-1,3-benzodiazole
77123
2-(2-chlorophenyl)-1H-benzimidazole
C1=CC=C(C(=C1)C2=NC3=CC=CC=C3N2)Cl
This item is not returnable. View return policy
3574-96-7
C13H9ClN2
228.679
MFCD00051632
OILIYWFQRJOPAI-UHFFFAOYSA-N
chlorfenazole, 2-2-chlorophenyl benzimidazole, 2-2-chlorophenyl-1h-benzimidazole, chlorfenazole iso, 1h-benzimidazole, 2-2-chlorophenyl, 2-2-chlorophenyl-1h-benzoimidazole, 2-2-chlorophenyl-1h-benzo d imidazole, unii-rt668w2svp, 2-o-chlorophenyl benzimidazole, 2-2-chlorophenyl-1h-1,3-benzodiazole
77123
2-(2-chlorophenyl)-1H-benzimidazole
C1=CC=C(C(=C1)C2=NC3=CC=CC=C3N2)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 232°C to 236°C
Quantity 5 g
Beilstein 173013
Formula Weight 228.68
Percent Purity 98%
Chemical Name or Material 2-(2-Chlorophenyl)benzimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'