Läs mer
2-[[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]amino]-3-nitro-benzoic Acid Methyl Ester, TRC
Produktkod. 30351334
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30351334 10 mg
1 options
Produktkod. 30351334 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98832010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]amino]-3-nitro-benzoic Acid Methyl Ester
CAS 139481-28-0
Molekylformel C22 H17 N3 O4
Kvantitet 10 mg
Synonym Methyl 2-[[(2'-Cyanobiphenyl-4-yl)methyl]amino]-3-nitrobenzoate, Candesartan nitro impurity
InChI formel InChI=1S/C22H17N3O4/c1-29-22(26)19-7-4-8-20(25(27)28)21(19)24-14-15-9-11-16(12-10-15)18-6-3-2-5-17(18)13-23/h2-12,24H,14H2,1H3
LEDER COC(=O)c1cccc(c1NCc2ccc(cc2)c3ccccc3C#N)[N+](=O)[O-]
IUPAC-namn methyl 2-[[4-(2-cyanophenyl)phenyl]methylamino]-3-nitrobenzoate
Molekylvikt (g/mol) 387.39
Formel vikt 387.1219