Läs mer
2,2-Dichloro-N-(2,6-dimethylphenyl)acetamide, Mikromol™
Produktkod. 30049494
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049494 25 mg
17944166 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049494 Leverantör Mikromol Leverantörsnummer MM0102.180025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2,2-Dichloro-N-(2,6-dimethylphenyl)acetamide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,2-Dichloro-N-(2,6-dimethylphenyl)acetamide
CAS 39084-88-3
Analyt- eller komponentnamn 2,2-Dichloro-N-(2,6-dimethylphenyl)acetamide
Frakt skick Room Temperature
Molekylformel C10 H11 Cl2 N O
Certifikat och efterlevnad ISO 17025
Synonym 2,2-Dichloro-N-(2,6-dimethylphenyl)acetamide, N-(2,6-Dimethylphenyl)-2,2-dichloroacetamide, NSC 37288
InChI formel InChI=1S/C10H11Cl2NO/c1-6-4-3-5-7(2)8(6)13-10(14)9(11)12/h3-5,9H,1-2H3,(H,13,14)
LEDER Cc1cccc(C)c1NC(=O)C(Cl)Cl
IUPAC-namn 2,2-dichloro-N-(2,6-dimethylphenyl)acetamide
Molekylvikt (g/mol) 232.11
Kvantitet 25 mg
Formel vikt 231.0218 g/mol