Läs mer
2,2-Dichloropropanoic Acid (>90%), TRC
Produktkod. 30370653
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30370653 500 mg
1 options
Produktkod. 30370653 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD436810500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dalapon
CAS 75-99-0
Molekylformel C3 H4 Cl2 O2
Kvantitet 500 mg
Synonym Propionic acid, 2,2-dichloro- (8CI), 2,2-Dichloropropanoic acid, 2,2-DPA, 2,2-Dichloropropionic acid, Alatex, Basinex P, DPA, Dalapon, Dalascam, NSC 56352, Proprop, S 1315, S 95, S 95 (herbicide), Tripon, α,α-Dichloropropanoic acid, α,α-Dichloropropionic acid
InChI formel InChI=1S/C3H4Cl2O2/c1-3(4,5)2(6)7/h1H3,(H,6,7)
LEDER CC(Cl)(Cl)C(=O)O
IUPAC-namn 2,2-dichloropropanoic acid
Molekylvikt (g/mol) 142.97
Formel vikt 141.9588