Läs mer
2-(2-Diethylaminoethoxy)ethanol, TRC
Produktkod. 30137940
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137940 250 mg
30134497 100 mg
30009730 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137940 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD443615250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 140-82-9
Molekylformel C8 H19 N O2
Molekylvikt (g/mol) 161.24
Synonym 2-[2-(Diethylamino)ethoxy]ethanol, Oxeladin Hydrogen Citrate Imp. A (EP)
IUPAC-namn 2-[2-(diethylamino)ethoxy]ethanol
LEDER CCN(CC)CCOCCO
Specifikationer
Kemiskt namn eller material 2-[2-(Diethylamino)ethoxy]ethanol
Anteckningar om renhetsgrad GC
Kvantitet 250 mg
InChI formel InChI=1S/C8H19NO2/c1-3-9(4-2)5-7-11-8-6-10/h10H,3-8H2,1-2H3
Formel vikt 161.1416
Procent renhet >95%