Learn More
2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid, 97%, Thermo Scientific™
Catalog Number 15189323
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
656-46-2
C8H4F2O4
202.12
VJLDRFCNFNQTTH-UHFFFAOYSA-N
2,2-difluorobenzo d 1,3 dioxole-5-carboxylic acid, 2,2-difluoro-2h-1,3-benzodioxole-5-carboxylic acid, 2,2-difluorobenzodioxole-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxole-5-carboxylic acid, 3,4-difluoromethylenedioxy benzoic acid, acmc-209nrl, 5-carboxy-2,2-difluoro-1,3-benzodioxole, 2,2-difluorobenzo-1,3dioxole-5-carboxylic acid, 2,2-difluorobenzo 1,3 dioxol-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxol-5-carboxylic acid
608772
2,2-difluoro-1,3-benzodioxole-5-carboxylic acid
C1=CC2=C(C=C1C(=O)O)OC(O2)(F)F
656-46-2
C8H4F2O4
202.12
VJLDRFCNFNQTTH-UHFFFAOYSA-N
2,2-difluorobenzo d 1,3 dioxole-5-carboxylic acid, 2,2-difluoro-2h-1,3-benzodioxole-5-carboxylic acid, 2,2-difluorobenzodioxole-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxole-5-carboxylic acid, 3,4-difluoromethylenedioxy benzoic acid, acmc-209nrl, 5-carboxy-2,2-difluoro-1,3-benzodioxole, 2,2-difluorobenzo-1,3dioxole-5-carboxylic acid, 2,2-difluorobenzo 1,3 dioxol-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxol-5-carboxylic acid
608772
2,2-difluoro-1,3-benzodioxole-5-carboxylic acid
C1=CC2=C(C=C1C(=O)O)OC(O2)(F)F
Melting Point 153°C to 157°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 202.12
Percent Purity 97%
Chemical Name or Material 2, 2-Difluoro-1, 3-benzodioxole-5-carboxylic acid
compliance-icons
Product Identifier
  • 2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.