Läs mer
2’,2’-Difluoro-2’-deoxyuridine, TRC
Produktkod. 30011151
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011151 5 mg
30007282 10 mg
30005474 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011151 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4457405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 114248-23-6
Molekylformel C9 H10 F2 N2 O5
Molekylvikt (g/mol) 264.18
Synonym Uridine, 2'-deoxy-2',2'-difluoro-, 2'-Deoxy-2',2'-difluorouridine, 2',2'-Difluoro-2'-deoxyuridine, 2',2'-Difluorodeoxyuridine, 1-(2-Deoxy-2,2-difluoro-β-D-erythro-pentofuranosyl)pyrimidin-2,4(1H,3H)-dione
IUPAC-namn 1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
LEDER OC[C@H]1O[C@@H](N2C=CC(=O)NC2=O)C(F)(F)[C@@H]1O
Specifikationer
Kemiskt namn eller material 2',2'-difluoro-2'-deoxyuridine
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C9H10F2N2O5/c10-9(11)6(16)4(3-14)18-7(9)13-2-1-5(15)12-8(13)17/h1-2,4,6-7,14,16H,3H2,(H,12,15,17)/t4-,6-,7-/m1/s1
Formel vikt 264.0558
Procent renhet >95%