Läs mer
2,2'-Dihydroxy-4,4'-dimethoxybenzophenone-5,5'-disulfonic Acid Disodium Salt, (Technical Grade), TRC
Produktkod. 30373597
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30373597 1 mg
1 options
Produktkod. 30373597 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4940781MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,2'-Dihydroxy-4,4'-dimethoxybenzophenone-5,5'-disulfonic Acid Disodium Salt, (Technical Grade)
CAS 76656-36-5
Molekylformel C15H12Na2O11S2
Kvantitet 1 mg
Synonym Uvinul DS 49, Uvinul 3048, Benzophenone 9
InChI formel InChI=1S/C15H14O11S2.2Na/c1-25-11-5-9(16)7(3-13(11)27(19,20)21)15(18)8-4-14(28(22,23)24)12(26-2)6-10(8)17;;/h3-6,16-17H,1-2H3,(H,19,20,21)(H,22,23,24);;/q;2*+1/p-2
LEDER [Na+].[Na+].COc1cc(O)c(cc1S(=O)(=O)[O-])C(=O)c2cc(c(OC)cc2O)S(=O)(=O)[O-]
IUPAC-namn disodium;4-hydroxy-5-(2-hydroxy-4-methoxy-5-sulfonatobenzoyl)-2-methoxybenzenesulfonate
Molekylvikt (g/mol) 478.36
Formel vikt 477.9616