Learn More
2,2-Dimethyl-1,3-dioxan-5-one, tech. 90%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
2,2-Dimethyl-1,3-dioxan-5-one is used in the preparation of 2,2-dimethyl-[1,3]dioxan-5-ol by reducing with lithium aluminum hydride. It is also used in the annulation of bete-(hetero)aryl-alfa-nitro-alfa,beta-enals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2-Dimethyl-1,3-dioxan-5-one is used in the preparation of 2,2-dimethyl-[1,3]dioxan-5-ol by reducing with lithium aluminum hydride. It is also used in the annulation of bete-(hetero)aryl-alfa-nitro-alfa,beta-enals.
Solubility
Miscible with water and most organic solvents.
Notes
Incompatible with oxidizing agents. Air, moisture and heat sensitive.
2,2-Dimethyl-1,3-dioxan-5-one is used in the preparation of 2,2-dimethyl-[1,3]dioxan-5-ol by reducing with lithium aluminum hydride. It is also used in the annulation of bete-(hetero)aryl-alfa-nitro-alfa,beta-enals.
Solubility
Miscible with water and most organic solvents.
Notes
Incompatible with oxidizing agents. Air, moisture and heat sensitive.
Specifications
Boiling Point | 40°C to 46°C (14 mmHg) |
Refractive Index | 1.4315 |
Quantity | 250 mg |
Beilstein | 3061933 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Miscible with water and most organic solvents. |
Formula Weight | 130.15 |
Percent Purity | 90% |
Grade | Technical |
Chemical Name or Material | 2,2-Dimethyl-1,3-dioxan-5-one |