missing translation for 'onlineSavingsMsg'
Läs mer

2,2-Dimethyl-1,3-dioxane-5-carboxylic Acid Methyl Ester, TRC

Produktkod. 30378544
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30378544 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30378544 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD472620100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2,2-Dimethyl-1,3-dioxane-5-carboxylic Acid Methyl Ester
CAS 155818-14-7
Molekylformel C8 H14 O4
Kvantitet 100 mg
InChI formel InChI=1S/C8H14O4/c1-8(2)11-4-6(5-12-8)7(9)10-3/h6H,4-5H2,1-3H3
LEDER COC(=O)C1COC(C)(C)OC1
IUPAC-namn methyl 2,2-dimethyl-1,3-dioxane-5-carboxylate
Molekylvikt (g/mol) 174.19
Formel vikt 174.0892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.