missing translation for 'onlineSavingsMsg'
Läs mer

2,2-Dimethyl-1,3-dioxolane-4-carboxylic acid, TRC

Produktkod. 30375723
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30375723 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30375723 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD48005810MG

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 2,2-Dimethyl-1,3-dioxolane-4-carboxylic acid
CAS 5736-06-1
Molekylformel C6 H10 O4
Kvantitet 10 mg
InChI formel InChI=1S/C6H10O4/c1-6(2)9-3-4(10-6)5(7)8/h4H,3H2,1-2H3,(H,7,8)
LEDER CC1(C)OCC(O1)C(=O)O
IUPAC-namn 2,2-dimethyl-1,3-dioxolane-4-carboxylic acid
Molekylvikt (g/mol) 146.141
Formel vikt 146.058
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.