Läs mer
2,2-Dimethylacetoacetic Acid Ethyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2,2- Dimethyl-acetoacetic Acid Ethyl Ester |
| CAS | 597-04-6 |
| Molekylformel | C8 H14 O3 |
| Kvantitet | 500 mg |
| Synonym | Butanoic acid, 2,2-dimethyl-3-oxo-, ethyl ester, Acetoacetic acid, 2,2-dimethyl-, ethyl ester (6CI,7CI,8CI), 2,2-Dimethyl-3-oxobutanoic acid ethyl ester, 2,2-Dimethyl-3-oxobutyric acid ethyl ester, 2-Acetyl-2-(ethoxycarbonyl)propane, Ethyl 2,2-dimethyl-3-oxobutanoate, Ethyl 2,2-dimethyl-3-oxobutyrate, Ethyl 2,2-dimethylacetoacetate, Ethyl 2-acetyl-2-methylpropionate, Ethyl acetyldimethylacetate, Ethyl α,α-dimethylacetoacetate, NSC 29039 |
| InChI formel | InChI=1S/C8H14O3/c1-5-11-7(10)8(3,4)6(2)9/h5H2,1-4H3 |
| LEDER | CCOC(=O)C(C)(C)C(=O)C |
| IUPAC-namn | ethyl 2,2-dimethyl-3-oxobutanoate |
| Molekylvikt (g/mol) | 158.2 |
| Formel vikt | 158.0943 |