Läs mer
2-{[2-(Dimethylamino)ethyl]methylamino}ethanol, TRC
Produktkod. 30370787
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30370787 10 g
1 options
Produktkod. 30370787 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47408010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-{[2-(Dimethylamino)ethyl]methylamino}ethanol
CAS 2212-32-0
Molekylformel C7H18N2O
Kvantitet 10 g
Synonym 2-(2-N,N-Dimethylaminoethyl-N-methylamino)ethanol, 2-[[2-(Dimethylamino)ethyl]methylamino]ethanol, Dabco T, Jeffcat Z 110, Kaolizer 28, N,N,N'-Trimethyl-N'-(2-hydroxyethyl)ethylenediamine, N,N,N'-Trimethyl-N'-(hydroxyethyl)ethylenediamine, N,N,N'-Trimethylaminoethyl Ethanolamine, N-(2-Dimethylaminoethyl)-N-methylethanolamine, N-(Dimethylaminoethyl)-N-methylethanolamine, RX 5, Toyocat RX 5, Z 110
InChI formel InChI=1S/C7H18N2O/c1-8(2)4-5-9(3)6-7-10/h10H,4-7H2,1-3H3
LEDER OCCN(CCN(C)C)C
IUPAC-namn 2-[2-(dimethylamino)ethyl-methylamino]ethanol
Molekylvikt (g/mol) 146.23
Formel vikt 146.1419