Läs mer
2,2-dimethylhexanal, TRC
Produktkod. 30351925
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351925 1 g
30360994 500 mg
30349061 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30351925 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2067161G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 996-12-3
Molekylformel C8H16O
Molekylvikt (g/mol) 128.21
IUPAC-namn 2,2-dimethylhexanal
LEDER CCCCC(C)(C)C=O
Specifikationer
Kemiskt namn eller material 2,2-dimethylhexanal
Kvantitet 1 g
InChI formel InChI=1S/C8H16O/c1-4-5-6-8(2,3)7-9/h7H,4-6H2,1-3H3
Formel vikt 128.1201