missing translation for 'onlineSavingsMsg'
Läs mer

2,2'-Dithiobis[2-methyl-1-propanol], TRC

Produktkod. 30368947
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368947 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30368947 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4941701G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 2,2'-Dithiobis[2-methyl-1-propanol]
CAS 132182-01-5
Molekylformel C8H18O2S2
Kvantitet 1 g
Synonym 2,2'-Disulfanediylbis(2-methylpropan-1-ol)
InChI formel InChI=1S/C8H18O2S2/c1-7(2,5-9)11-12-8(3,4)6-10/h9-10H,5-6H2,1-4H3
LEDER OCC(SSC(C)(C)CO)(C)C
IUPAC-namn 2,2'-disulfanediylbis(2-methylpropan-1-ol)
Molekylvikt (g/mol) 210.36
Formel vikt 210.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.