Läs mer
2,2’-(Ethanediyldiimino)bis-butanoic Acid(Mixture of Diastereomers), TRC
Produktkod. 30376760
Change view
Kvantitet:
100 mg
500 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30376760 5 g
30376403 500 mg
30375624 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30376760 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE8903405G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 498-17-9
Molekylformel C10 H20 N2 O4
Molekylvikt (g/mol) 232.28
Synonym 2,2'-(Ethylenediimino)di-butyric Acid, Ethylenediamine-N,N'-di-α-butyric Acid
IUPAC-namn 2-[2-(1-carboxypropylamino)ethylamino]butanoic acid
LEDER CCC(NCCNC(CC)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material 2,2'-(Ethanediyldiimino)bis-butanoic acid
Kvantitet 5 g
InChI formel InChI=1S/C10H20N2O4/c1-3-7(9(13)14)11-5-6-12-8(4-2)10(15)16/h7-8,11-12H,3-6H2,1-2H3,(H,13,14)(H,15,16)
Formel vikt 232.1423