missing translation for 'onlineSavingsMsg'
Läs mer

2-(2-Ethoxyphenoxy)ethyl Bromide, TRC

Produktkod. 30380991
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30380991 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30380991 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE892663250MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 1-(2-Bromoethoxy)-2-ethoxy-benzene
CAS 3259-03-8
Molekylformel C10 H13 Br O2
Kvantitet 250 mg
Synonym 1-(2-Bromoethoxy)-2-ethoxybenzene, Benzene, 1-(2-bromoethoxy)-2-ethoxy- (7CI,8CI,9CI), 2-(o-Ethoxyphenoxy)ethyl bromide, 2-(2-Ethoxyphenoxy)ethyl Bromide
InChI formel InChI=1S/C10H13BrO2/c1-2-12-9-5-3-4-6-10(9)13-8-7-11/h3-6H,2,7-8H2,1H3
LEDER CCOc1ccccc1OCCBr
IUPAC-namn 1-(2-bromoethoxy)-2-ethoxybenzene
Molekylvikt (g/mol) 245.11
Formel vikt 244.0099
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.