Läs mer
2-(2-Ethoxyphenyl)-5,7-dimethylimidazo[5,1-f][1,2,4]triazin-4(1H)-one, TRC
Produktkod. 30374182
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374182 5 mg
30371208 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30374182 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE6795855MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 224789-20-2
Molekylformel C15 H22 N4 O3
Molekylvikt (g/mol) 306.36
IUPAC-namn 3-methyl-7-(5-oxohexyl)-1-propylpurine-2,6-dione
LEDER CCCN1C(=O)N(C)c2ncn(CCCCC(=O)C)c2C1=O
Specifikationer
Kemiskt namn eller material 2-(2-Ethoxyphenyl)-5,7-dimethylimidazo[5,1-f][1,2,4]triazin-4(1H)-one
Kvantitet 5 mg
InChI formel InChI=1S/C15H22N4O3/c1-4-8-19-14(21)12-13(17(3)15(19)22)16-10-18(12)9-6-5-7-11(2)20/h10H,4-9H2,1-3H3
Formel vikt 306.169