Läs mer
2-[2-[(EZ)-1,2-Diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine, Mikromol™
Produktkod. 18094040
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18094040 100 mg
18080850 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18094040 Leverantör Mikromol Leverantörsnummer MM0053.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[2-[(EZ)-1,2-Diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[2-[(EZ)-1,2-Diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine
CAS 2734357-02-7
Analyt- eller komponentnamn 2-[2-[(EZ)-1,2-Diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine
Frakt skick Room Temperature
Molekylformel C26 H29 N O
Certifikat och efterlevnad ISO 9001
Synonym Tamoxifen Citrate Imp. E (EP), Tamoxifen Imp. E (EP), 2-[2-[(EZ)-1,2-Diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine, Tamoxifen Citrate Impurity E, Tamoxifen Impurity E
InChI formel InChI=1S/C17H14N4O4/c1-25-17(24)21-16-19-13-7-4-11(8-14(13)20-16)15(23)10-2-5-12(6-3-10)18-9-22/h2-9H,1H3,(H,18,22)(H2,19,20,21,24)
LEDER CCC(=C(c1ccccc1)c2ccccc2OCCN(C)C)c3ccccc3
IUPAC-namn 2-[2-(1,2-diphenylbut-1-enyl)phenoxy]-N,N-dimethylethanamine
Molekylvikt (g/mol) 371.51
Kvantitet 100 mg
Formel vikt 371.2249 g/mol