Learn More
Alfa Aesar™ 2-(2-Fluorophenyl)-5-methylbenzimidazole, 95%
Brand:  Alfa Aesar™ H64160.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15481229
  • 3114.20 SEK / 1g
Estimated Shipment date
from Supplier 07-05-2018
Add to basket
Specifications
Chemical Identifiers
2-(2-Fluorophenyl)-5-methylbenzimidazole
226.25
MFCD18914843
Specifications
2-(2-Fluorophenyl)-5-methylbenzimidazole
Solid
226.25
1g
95%
MFCD18914843