missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Fluorophenyl)-5-nitrobenzimidazole, 95%

Brand:  Alfa Aesar™ H64660.03

Additional Details : CAS Number : 348-33-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15451949

  • 3114.20 SEK / 1g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

2-(2-Fluorophenyl)-5-nitrobenzimidazole
348-33-4
257.22
MFCD13812381

Specifications

2-(2-Fluorophenyl)-5-nitrobenzimidazole
348-33-4
95%
MFCD13812381
1g
Solid
257.22
Videos
SDS