Läs mer
2,2’-Iminobis(N-(2,6-Dimethylphenyl)acetiamide, TRC
Produktkod. 30010439
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010439 100 mg
30004301 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010439 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI400100100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 745798-07-6
Molekylformel C20H25N3O2
Molekylvikt (g/mol) 339.43
Synonym 2,2'-Iminobis(N-(2,6-Dimethylphenyl)acetiamide
IUPAC-namn 2-[[2-(2,6-dimethylanilino)-2-oxoethyl]amino]-N-(2,6-dimethylphenyl)acetamide
LEDER O=C(NC1=C(C)C=CC=C1C)CNCC(NC2=C(C)C=CC=C2C)=O
Specifikationer
Kemiskt namn eller material USP Lidocaine Impurity E
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C20H25N3O2/c1-13-7-5-8-14(2)19(13)22-17(24)11-21-12-18(25)23-20-15(3)9-6-10-16(20)4/h5-10,21H,11-12H2,1-4H3,(H,22,24)(H,23,25)
Formel vikt 339.1947
Procent renhet >95%