Läs mer
2-[(2-Methoxyacetyl)(2-methoxy-1-methyl-2-oxoethyl)amino]-3-methylbenzoic Acid, TRC
Produktkod. 30476653
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30476653 25 mg
1 options
Produktkod. 30476653 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM91225025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Metalaxyl Metabolite CGA 108905
CAS 96258-91-2
Molekylformel C15 H19 N O6
Kvantitet 25 mg
Synonym 2-[(2-Methoxyacetyl)(2-methoxy-1-methyl-2-oxoethyl)amino]-3-methylbenzoic Acid, Metalaxyl Metabolite CGA 108905','2-[(2-Methoxyacetyl)(2-methoxy-1-methyl-2-oxoethyl)amino]-3-methylbenzoic acid (ACI), Benzoic acid','2-[(methoxyacetyl)(2-methoxy-1-methyl-2-oxoethyl)amino]-3-methyl- (9CI), Benzoic acid','2-[(methoxyacetyl)(2-methoxy-1-meth
InChI formel InChI=1S/C15H19NO6/c1-9-6-5-7-11(14(18)19)13(9)16(12(17)8-21-3)10(2)15(20)22-4/h5-7,10H,8H2,1-4H3,(H,18,19)
LEDER COCC(=O)N(C(C)C(=O)OC)c1c(C)cccc1C(=O)O
IUPAC-namn 2-[(2-methoxyacetyl)-(1-methoxy-1-oxopropan-2-yl)amino]-3-methylbenzoic acid
Molekylvikt (g/mol) 309.315
Formel vikt 309.121