Läs mer
2-(2-Methoxyphenoxy)-1,3-propanediol, TRC
Produktkod. 30004165
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004165 50 mg
30007296 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004165 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM27163050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14007-09-1
Molekylformel C10 H14 O4
Molekylvikt (g/mol) 198.22
Synonym 2-(2-Methoxyphenoxy)propane-1,3-diol, Guaifenesin Imp. B (EP), B-Isomer of Guaifenesin, Isoguaifenesin
IUPAC-namn 2-(2-methoxyphenoxy)propane-1,3-diol
LEDER COc1ccccc1OC(CO)CO
Specifikationer
Kemiskt namn eller material 2-(2-Methoxyphenoxy)propane-1,3-diol
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C10H14O4/c1-13-9-4-2-3-5-10(9)14-8(6-11)7-12/h2-5,8,11-12H,6-7H2,1H3
Formel vikt 198.0892
Procent renhet >95%