Läs mer
2-(2-Methoxyphenoxy)acetaldehyde, TRC
Produktkod. 30471278
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471278 10 mg
30469217 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30471278 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26542010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18167-91-4
Molekylformel C9H10O3
Molekylvikt (g/mol) 166.18
Synonym (o-Methoxyphenoxy)acetaldehyde, Guaiacoxyacetaldehyde, O-(2-Oxoethyl)guaiacol
IUPAC-namn 2-(2-methoxyphenoxy)acetaldehyde
LEDER COc1ccccc1OCC=O
Specifikationer
Kemiskt namn eller material 2-(2-Methoxyphenoxy)acetaldehyde
Kvantitet 10 mg
InChI formel InChI=1S/C9H10O3/c1-11-8-4-2-3-5-9(8)12-7-6-10/h2-6H,7H2,1H3
Formel vikt 166.063