Läs mer
2-(2-Methoxyphenoxy)propane-1,3-diol (B-Isomer), Mikromol™
Produktkod. 30049251
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049251 100 mg
30049539 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049251 Leverantör Mikromol Leverantörsnummer MM0354.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(2-Methoxyphenoxy)propane-1,3-diol (B-Isomer)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(2-Methoxyphenoxy)propane-1,3-diol
CAS 14007-09-1
Analyt- eller komponentnamn 2-(2-Methoxyphenoxy)propane-1,3-diol
Frakt skick Room Temperature
Molekylformel C10 H14 O4
Certifikat och efterlevnad ISO 17025
Synonym Guaifenesin Imp. B (EP), 2-(2-Methoxyphenoxy)propane-1,3-diol, B-Isomer of Guaifenesin, Guaifenesin Impurity B
InChI formel InChI=1S/C10H14O4/c1-13-9-4-2-3-5-10(9)14-8(6-11)7-12/h2-5,8,11-12H,6-7H2,1H3
LEDER COc1ccccc1OC(CO)CO
IUPAC-namn 2-(2-methoxyphenoxy)propane-1,3-diol
Molekylvikt (g/mol) 198.22
Kvantitet 100 mg
Formel vikt 198.0892 g/mol