Learn More
Alfa Aesar™ 2-(2-Methoxyphenyl)-5-methylbenzimidazole, 95%
Brand:  Alfa Aesar™ H64063.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15471089
  • 11319.11 SEK / 5g
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
2-(2-Methoxyphenyl)-5-methylbenzimidazole
238.29
MFCD11849219
Specifications
2-(2-Methoxyphenyl)-5-methylbenzimidazole
95%
MFCD11849219
Solid
238.29
5g