Läs mer
2,2'-Methylenebis(6-tert-butyl-p-cresol), TRC
Produktkod. 30135555
Change view
Kvantitet:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135555 100 g
30007584 5 g
30004558 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135555 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM303938100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 119-47-1
Molekylformel C23 H32 O2
Molekylvikt (g/mol) 340.5
Synonym p-Cresol, 2,2'-methylenebis[6-tert-butyl- (8CI), 2,2'-Methylenebis[6-(1,1-dimethylethyl)-4-methylphenol], 2,2- Methylene bis(4-methyl-6-tert-butyl-phenol), 2,2'-Bis(4-methyl-6-tert-butylphenol)methane, 2,2'-Methylenebis(4-methyl-6-tert-butylphenol), 2,2'-Methylenebis(6-tert-butyl-4-methylphenol), 2,2'-Methylenebis(6-tert-butyl-p-cresol), 2,2'-Methylenebis[4-methyl-6-t-butylphenol], 2,2'-Methylenebis[4-methyl-6-tert-butylphenol], 2,2'-Methylenebis[6-tert-butyl-p-cresol], 2,2'-Methylidenebis(6-tert-butyl-4-methylphenol), 3,3'-Di-tert-butyl-2,2'-dihydroxy-5,5'-dimethyldiphenylmethane, 6,6-Methylenebis(2-tert-butyl-4-methylphenol), 6,6'-Di-tert-butyl-4,4'-dimethyl-2,2'-methylenediphenol, 6,6'-Methylenebis(2-tert-butyl-4-methylphenol), A 02246, AO 1, AO 1 (antioxidant), AO 2246, AO 235, Additin RC 7115, Advastab 405, Agidol 2, Akrochem AO 235, Antage W 400, Anti Ox, Antioxidant 1, Antioxidant 2246, Antioxidant BKF, Antioxidant MBP, Antioxidant NG 2246, Antioxidant OMB, BKF, Baynox Plus, Bis(2-hydroxy-3-tert-butyl-5-methylphenyl)methane, Bis(2-hydroxy-5-methyl-3-tert-butylphenyl)methane, Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)methane, Bis(5-methyl-3-tert-butyl-2-hydroxyphenyl)methane, Bis(6-hydroxy-3-methyl-5-tert-butylphenyl)methane, Bisalkofen BP, CAO 14, CAO 5, Calco 2246, Catolin 14, Chemanox 21, Cyanox 2246, GERI-BP 002A, Ionol 46, Irganox 2246, Lederle 2246, Lowinox 22M46, Lowinox 22M48, MBP 5, MDP, NG 2246, NS 6, NSC 7781, Naftonox 22M46, Noclizer NS 6, Nocrac NS 6, Nonflex MBP, Nonflex MPP, Ongrostab 2246, Plastanox 2246, Product 2246, Santowhite PC, Sumilizer MDP, Sumilizer MDP 9, Sumilizer MDP-S, Vanox MBPC, Vulkanox BKF, Vulkanox BPH, Yoshinox 2246G, Yoshinox 2246R
IUPAC-namn 2-tert-butyl-6-[(3-tert-butyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
LEDER Cc1cc(Cc2cc(C)cc(c2O)C(C)(C)C)c(O)c(c1)C(C)(C)C
Specifikationer
Kemiskt namn eller material 2,2'-Methylene-bis(6-tert-butyl-4-methylphenol)
Kvantitet 100 g
InChI formel InChI=1S/C23H32O2/c1-14-9-16(20(24)18(11-14)22(3,4)5)13-17-10-15(2)12-19(21(17)25)23(6,7)8/h9-12,24-25H,13H2,1-8H3
Formel vikt 340.2402