Läs mer
2,2'-Methylenedianiline, TRC
Produktkod. 30476234
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30476234 50 mg
1 options
Produktkod. 30476234 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM23669050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,2'-Methylenedianiline
CAS 6582-52-1
Molekylformel C13 H14 N2
Kvantitet 50 mg
Synonym Aniline, 2,2'-methylenedi- (7CI,8CI), 2,2'-Methylenebis[benzenamine], 2,2'-Bis(aminophenyl)methane, 2,2'-Diaminodiphenylmethane, 2,2'-Diphenylmethanediamine, 2,2'-Methylenebisaniline, 2,2'-Methylenedianiline, 2,2'-Methylenedibenzenamine, 2,2'-Methylenediphenylamine, 2-[(2-Aminophenyl)methyl]aniline, o,o'-Bis(aminophenyl)methane, o,o'-Diaminodiphenylmethane, o,o'-Methylenedianiline
InChI formel InChI=1S/C13H14N2/c14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)15/h1-8H,9,14-15H2
LEDER Nc1ccccc1Cc2ccccc2N
IUPAC-namn 2-[(2-aminophenyl)methyl]aniline
Molekylvikt (g/mol) 198.26
Formel vikt 198.1157