Läs mer
2,2'-Sulfonyldiethanol, TRC
Produktkod. 30418440
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30418440 1 g
1 options
Produktkod. 30418440 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6994201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Thiodiglycol sulfone
CAS 2580-77-0
Molekylformel C4 H10 O4 S
Kvantitet 1 g
Synonym Ethanol','2,2'-sulfonylbis-, Ethanol','2,2'-sulfonyldi- (6CI,8CI), 2,2'-Sulfonylbis[ethanol], 2,2'-Sulfonyldi[ethanol], 2-(2-Hydroxyethanesulfonyl)ethan-1-ol, 2-(2-Hydroxyethylsulfonyl)ethanol, BHES 50, Bis(2-hydroxyethyl) sulfone, Bis(β-hydroxyethyl) sulfone, NSC 32716, NSC 50845, β,β'-Dihydroxyethyl sulfone, β-Hydroxyethyl sulfone
InChI formel InChI=1S/C4H10O4S/c5-1-3-9(7,8)4-2-6/h5-6H,1-4H2
LEDER OCCS(=O)(=O)CCO
IUPAC-namn 2-(2-hydroxyethylsulfonyl)ethanol
Molekylvikt (g/mol) 154.185
Formel vikt 154.03