Learn More
2-(2-Thienyl)pyrrolidine Hydrochloride, TRC
Product Code. 30454813
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454813 50 mg 50mg
30455122 100 mg 100mg
30456357 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30454813 Supplier Toronto Research Chemicals Supplier No. TRCT77593550MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(2-Thienyl)pyrrolidine Hydrochloride
Molecular Formula C8 H11 N S . H Cl
Quantity 50 mg
InChI Formula InChI=1S/C8H11NS.ClH/c1-3-7(9-5-1)8-4-2-6-10-8;/h2,4,6-7,9H,1,3,5H2;1H
SMILES Cl.C1CNC(C1)c2cccs2
IUPAC Name 2-thiophen-2-ylpyrrolidine;hydrochloride
Molecular Weight (g/mol) 189.706
Formula Weight 189.038