Läs mer
2'-(2H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-carboxylic Acid, TRC
Produktkod. 30006263
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006263 100 mg
17941775 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006263 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT308515100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 164265-78-5
Molekylformel C14 H10 N4 O2
Molekylvikt (g/mol) 266.25
Synonym [1,1'-Biphenyl]-4-carboxylic acid, 2'-(2H-tetrazol-5-yl)-, [1,1'-Biphenyl]-4-carboxylic acid, 2'-(1H-tetrazol-5-yl)- (9CI), 2'-(2H-Tetrazol-5-yl)[1,1'-biphenyl]-4-carboxylic acid, Valsartanic Acid
IUPAC-namn 4-[2-(1H-tetrazol-5-yl)phenyl]benzoic acid
LEDER OC(=O)c1ccc(cc1)c2ccccc2c3nnn[nH]3
Specifikationer
Kemiskt namn eller material Valsartanic acid
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C14H10N4O2/c19-14(20)10-7-5-9(6-8-10)11-3-1-2-4-12(11)13-15-17-18-16-13/h1-8H,(H,19,20)(H,15,16,17,18)
Formel vikt 266.0804
Procent renhet >95%