Learn More
2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride, Thermo Scientific Chemicals
Product Code. 15490407
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490407 5 mg 5mg
15400417 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15490407 Supplier Thermo Scientific Alfa Aesar Supplier No. J64981.#0
 Restricted Item
Beräknad leverans: 14-12-2026
Add to basket
CAS: 418788-90-6 | Molecular Formula: C11H16Cl2NO2 | Molecular Weight: 265.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 418788-90-6
Molecular Formula C11H16Cl2NO2
Molecular Weight (g/mol) 265.15
MDL Number MFCD06410993
InChI Key QVEIRCZEBQRCTR-UHFFFAOYSA-O
Synonym 2,3-dcpe hydrochloride, 2-3-2,3-dichlorophenoxy propyl amino ethanol hydrochloride, c11h15cl2no2.hcl, 2-3-2,3-dichlorophenoxy propylamino ethanol hydrochloride
PubChem CID 52913859
IUPAC Name 2-[3-(2,3-dichlorophenoxy)propylamino]ethanol;hydrochloride
SMILES OCC[NH2+]CCCOC1=CC=CC(Cl)=C1Cl
Specifications
Color White
Quantity 5 mg
Solubility Information Soluble in water at ∽25mg/ml; Also soluble in ethanol,DMSO or DMF at ∽20mg/ml; Soluble in PBS,pH 7;2 at ∽10mg/ml.
Formula Weight 300.61
Physical Form Powder
Chemical Name or Material 2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride