Läs mer
[2-[[3-(2-Chlorobenzoyl)-5-ethyl-2-thienyl]amino]-2-oxoethyl]carbamic Acid Benzyl Ester, TRC
Produktkod. 30352433
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30352433 100 mg
1 options
Produktkod. 30352433 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC364970100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material [2-[(3-(2-Chlorobenzoyl)-5-ethyl-2-thienyl)amino]-2-oxoethyl]carbamic Acid Benzyl Ester
CAS 50508-73-1
Molekylformel C23H21ClN2O4S
Kvantitet 100 mg
Synonym [2-[[3-(2-Chlorobenzoyl)-5-ethyl-2-thienyl]amino]-2-oxoethyl]carbamic Acid Phenylmethyl Ester
InChI formel InChI=1S/C23H21ClN2O4S/c1-2-16-12-18(21(28)17-10-6-7-11-19(17)24)22(31-16)26-20(27)13-25-23(29)30-14-15-8-4-3-5-9-15/h3-12H,2,13-14H2,1H3,(H,25,29)(H,26,27)
LEDER CCC1=CC(=C(S1)NC(=O)CNC(=O)OCC2=CC=CC=C2)C(=O)C3=CC=CC=C3Cl
IUPAC-namn benzyl N-[2-[[3-(2-chlorobenzoyl)-5-ethylthiophen-2-yl]amino]-2-oxoethyl]carbamate
Molekylvikt (g/mol) 456.94
Formel vikt 456.0911