Läs mer
2,3,3,3-Tetrafluoro-2-(1,1,2,2,3,3,3,heptafluoropropoxy)propanoic Acid (>90%), TRC
Produktkod. 30456611
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30456611 1 g
1 options
Produktkod. 30456611 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3106201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluoro-2-propoxypropanoic acid (PFPrOPrA)
CAS 13252-13-6
Molekylformel C6 H F11 O3
Kvantitet 1 g
Synonym Propanoic acid','2,3,3,3-tetrafluoro-2-(heptafluoropropoxy)- (9CI), Propionic acid','2,3,3,3-tetrafluoro-2-(heptafluoropropoxy)- (7CI,8CI), 2,3,3,3-Tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propanoic acid, (±)-Perfluoro(2-propoxypropionic) acid, 2,3,3,3-Tetrafluoro-2-(1,1,2,2,3,3,3,heptafluoropropoxy)propanoic acid, Perfluoro-2-(propyloxy)propionic acid, Perfluoro-2-methyl-3-oxahexanoic acid, Perfluoro-2-propoxypropanoic acid, Perfluoro-2-propoxypropionic acid, Perfluoro-α-propoxypropionic acid, Perfluoro-2-propoxypropanoic Acid (PFPrOPrA )','GenX
InChI formel InChI=1S/C6HF11O3/c7-2(1(18)19,4(10,11)12)20-6(16,17)3(8,9)5(13,14)15/h(H,18,19)
LEDER OC(=O)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F
IUPAC-namn 2,3,3,3-tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propanoic acid
Molekylvikt (g/mol) 330.05
Formel vikt 329.975