Läs mer
2,3,3,3-Tetrafluoropropanoic Acid, TRC
Produktkod. 30009295
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009295 1 g
30007824 100 mg
30012842 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009295 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3109881G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 359-49-9
Molekylformel C3H2O2F4
Molekylvikt (g/mol) 146.04
Synonym Propanoic acid, 2,3,3,3-tetrafluoro- (9CI, ACI), 2,3,3,3-Tetrafluoropropanoic acid (ACI), Propionic acid, 2,3,3,3-tetrafluoro- (6CI, 7CI, 8CI), 2,3,3,3-Tetrafluoropropionic acid, 2H-Perfluoropropionic Acid, Propanoic acid, 2,3,3,3-tetrafluoro- (9CI, ACI), 2,3,3,3-Tetrafluoropropanoic acid (ACI), Propionic acid, 2,3,3,3-tetrafluoro- (6CI, 7CI, 8CI), 2,3,3,3-Tetrafluoropropionic acid, 2H-Perfluoropropionic Acid
IUPAC-namn 2,3,3,3-tetrafluoropropanoic acid
LEDER OC(=O)C(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material 2,3,3,3-Tetrafluoropropanoic Acid
Anteckningar om renhetsgrad GC
Kvantitet 1 g
InChI formel InChI=1S/C3H2F4O2/c4-1(2(8)9)3(5,6)7/h1H,(H,8,9)
Formel vikt 145.9991
Procent renhet >95%