Läs mer
2,3,3',4,4',5',6-Heptabromodiphenyl Ether, TRC
Produktkod. 30136959
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136959 250 mg
30041064 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136959 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH281070250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 446255-30-7
Molekylformel C12 H3 Br7 O
Molekylvikt (g/mol) 722.48
Synonym Benzene, 1,2,3,5-tetrabromo-4-(3,4,5-tribromophenoxy)-, 1,2,3,5-Tetrabromo-4-(3,4,5-tribromophenoxy)benzene, 2,3,3',4,4',5',6-Heptabromodiphenyl ether, BDE 191, PBDE 191, PBDE No. 191
IUPAC-namn 1,2,3,5-tetrabromo-4-(3,4,5-tribromophenoxy)benzene
LEDER Brc1cc(Oc2c(Br)cc(Br)c(Br)c2Br)cc(Br)c1Br
Specifikationer
Kemiskt namn eller material PBDE 191 (2,3,3',4,4',5',6-Heptabromodiphenyl Ether)
Kvantitet 250 mg
InChI formel InChI=1S/C12H3Br7O/c13-5-1-4(2-6(14)9(5)17)20-12-8(16)3-7(15)10(18)11(12)19/h1-3H
Formel vikt 715.4468