Läs mer
2,3,3-Trichloro-2-propenesulfonic Acid Sodium Salt, TRC
Produktkod. 30134378
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134378 50 mg
30010381 10 mg
30008878 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134378 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT79790050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 65600-61-5
Molekylformel C3 H2 Cl3 O3 S . Na
Molekylvikt (g/mol) 247.46
Synonym 2-Propene-1-sulfonic acid, 2,3,3-trichloro-, sodium salt (1:1), 2-Propene-1-sulfonic acid, 2,3,3-trichloro-, sodium salt, 2,3,3-Trichloro-2-propene-1-sulfonic acid sodium
IUPAC-namn sodium;2,3,3-trichloroprop-2-ene-1-sulfonate
LEDER [Na+].[O-]S(=O)(=O)CC(=C(Cl)Cl)Cl
Specifikationer
Kemiskt namn eller material 2,3,3-Trichloro-2-propene-1-sulfonic acid sodium
Kvantitet 50 mg
InChI formel InChI=1S/C3H3Cl3O3S.Na/c4-2(3(5)6)1-10(7,8)9;/h1H2,(H,7,8,9);/q;+1/p-1
Formel vikt 245.8688