Läs mer
2,3,3-Trimethyl-5-carboxyindolenine, TRC
Produktkod. 30452826
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30452826 100 mg
30449564 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30452826 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT795970100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84100-84-5
Molekylformel C12H13NO2
Molekylvikt (g/mol) 203.24
Synonym 2,3,3-Trimethyl-3H-indole-5-carboxylic Acid
IUPAC-namn 2,3,3-trimethylindole-5-carboxylic acid
LEDER CC1=Nc2ccc(cc2C1(C)C)C(=O)O
Specifikationer
Kemiskt namn eller material 2,3,3-Trimethyl-5-carboxyindolenine
Kvantitet 100 mg
InChI formel InChI=1S/C12H13NO2/c1-7-12(2,3)9-6-8(11(14)15)4-5-10(9)13-7/h4-6H,1-3H3,(H,14,15)
Formel vikt 203.0946