Läs mer
2,3,4,5,6-Pentachlorobenzeneamine, TRC
Produktkod. 30412754
Change view
Kvantitet:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412754 50 g
30421485 100 g
30417222 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30412754 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP23792550G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 527-20-8
Molekylformel C6 H2 Cl5 N
Molekylvikt (g/mol) 265.35
Synonym Aniline, 2,3,4,5,6-pentachloro- (6CI,7CI,8CI), Aniline, pentachloro- (5CI), 2,3,4,5,6-Pentachlorobenzenamine, 2,3,4,5,6-Pentachloroaniline, NSC 49579, PCA, Pentachloroaminobenzene, Pentachloroaniline
IUPAC-namn 2,3,4,5,6-pentachloroaniline
LEDER Nc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl
Specifikationer
Kemiskt namn eller material Pentachloroaniline
Kvantitet 50 g
InChI formel InChI=1S/C6H2Cl5N/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H2
Formel vikt 262.863