Läs mer
2,3,4,5-Tetradehydro Alfuzosin Hydrochloride, TRC
Produktkod. 30011347
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011347 10 mg -
30009365 25 mg -
30007858 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011347 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29143010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 98902-29-5
Molekylformel C19 H23 N5 O4 . Cl H
Molekylvikt (g/mol) 421.88
Synonym 2-Furancarboxamide, N-[3-[(4-amino-6,7-dimethoxy-2-quinazolinyl)methylamino]propyl]-, hydrochloride (1:1), 2-Furancarboxamide, N-[3-[(4-amino-6,7-dimethoxy-2-quinazolinyl)methylamino]propyl]-, monohydrochloride (9CI), N-[3-[(4-Amino-6,7-dimethoxyquinazolin-2-yl)methylamino]propyl]furan-2-carboxamide hydrochloride, 2,3,4,5-Tetradehydroalfuzosin hydrochloride
IUPAC-namn N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]furan-2-carboxamide;hydrochloride
LEDER Cl.COc1cc2nc(nc(N)c2cc1OC)N(C)CCCNC(=O)c3occc3
Specifikationer
Kemiskt namn eller material 2,3,4,5-Tetradehydro Alfuzosin hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C19H23N5O4.ClH/c1-24(8-5-7-21-18(25)14-6-4-9-28-14)19-22-13-11-16(27-3)15(26-2)10-12(13)17(20)23-19;/h4,6,9-11H,5,7-8H2,1-3H3,(H,21,25)(H2,20,22,23);1H
Formel vikt 421.1517