Läs mer
2,3,4,5-Tetrahydro-3-(trifluoroacetyl)-1,5-methano-1H-3-benzazepine-7,8-diamine, TRC
Produktkod. 30134820
Change view
Kvantitet:
100 mg
2.5 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134820 2.5 g
30137817 5 g
30009534 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134820 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2959852.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 230615-69-7
Molekylformel C13 H14 F3 N3 O
Molekylvikt (g/mol) 285.265
Synonym Ethanone, 1-(7,8-diamino-1,2,4,5-tetrahydro-1,5-methano-3H-3-benzazepin-3-yl)-2,2,2-trifluoro-, 1,5-Methano-1H-3-benzazepine-7,8-diamine, 2,3,4,5-tetrahydro-3-(trifluoroacetyl)- (9CI), 1-(7,8-Diamino-1,2,4,5-tetrahydro-1,5-methano-3H-3-benzazepin-3-yl)-2,2,2-trifluoroethanone
IUPAC-namn 1-(4,5-diamino-10-azatricyclo[6.3.1.02,7]dodeca-2,4,6-trien-10-yl)-2,2,2-trifluoroethanone
LEDER Nc1cc2C3CC(CN(C3)C(=O)C(F)(F)F)c2cc1N
Specifikationer
Kemiskt namn eller material 2,3,4,5-Tetrahydro-3-(trifluoroacetyl)-1,5-methano-1H-3-benzazepine-7,8-diamine
Kvantitet 2.5 g
InChI formel InChI=1S/C13H14F3N3O/c14-13(15,16)12(20)19-4-6-1-7(5-19)9-3-11(18)10(17)2-8(6)9/h2-3,6-7H,1,4-5,17-18H2
Formel vikt 285.1089